Triphenylen-2-ylboronic acid 98%

Supplier: Ambeed
Warning

A119358-1G A119358-250MG A119358-5G
76983-146EA 57.01 USD
76983-146 76983-148 77889-822
Triphenylen-2-ylboronic acid 98%
Triphenylen-2-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₃BO₂
MDL Number: MFCD16660186
CAS Number: 654664-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR