(S)-Ethyl morpholine-3-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A119334-100MG A119334-1G A119334-250MG
76983-140EA 45.62 USD
76983-140 76983-142 76983-144
(S)-Ethyl morpholine-3-carboxylate hydrochloride 95%
(S)-Ethyl morpholine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₃
MW: 195.64 g/mol
MDL Number: MFCD12755500
CAS Number: 218594-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR