Gastrodin 98%

Supplier: Ambeed
Warning

A119310-100G A119310-100MG A119310-1G A119310-25G A119310-5G
77009-348EA 372 USD
77009-348 77009-350 77009-352 77009-354 77009-356
Gastrodin 98%
Gastrodin

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₇
MDL Number: MFCD00272169
CAS Number: 62499-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR