Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-2-(4-Aminocyclohexyl)acetic acid Ethyl ester hydrochloride, Ethyl 2-(trans-4-aminocyclohexyl)acetate hydrochloride
A119295-100MG
A119295-10G
A119295-250MG
A119295-25G
A119295-1G
A119295-5G
76995-716EA
15.92
USD
76995-716
76995-718
76995-720
76995-722
77571-150
77670-686
Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride 95%
Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₂·HCl MW: 221.73 g/mol |
MDL Number:
MFCD16660366 CAS Number: 76308-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...