(3-Bromo-6-chloropyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A119250-1G A119250-250MG A119250-5G
76983-134EA 119.89 USD
76983-134 76983-136 76983-138
(3-Bromo-6-chloropyridin-2-yl)methanol 97%
(3-Bromo-6-chloropyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.46 g/mol
MDL Number: MFCD11847140
CAS Number: 1227601-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR