(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene)diisopropyl dicarbonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate, Tenofovir Disoproxil
A119233-100G
A119233-100MG
A119233-10MG
A119233-1G
A119233-25G
A119233-50MG
A119233-5G
A119233-250MG
76995-702EA
517
USD
76995-702
76995-704
76995-706
76995-708
76995-710
76995-712
76995-714
77880-965
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene)diisopropyl dicarbonate 98%
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene)diisopropyl dicarbonate
Precaución: for Research Use Only
Formula:
C₁₉H₃₀N₅O₁₀P MW: 519.44 g/mol |
MDL Number:
MFCD00952920 CAS Number: 201341-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...