5-Fluoro-2-methoxy-4-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-methoxypyridine-4-boronic acid

A119227-1G A119227-250MG A119227-25G A119227-5G
77009-340EA 67.9 USD
77009-340 77009-342 77009-344 77009-346
5-Fluoro-2-methoxy-4-pyridineboronic acid 95%
5-Fluoro-2-methoxy-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₃
MW: 170.9300 g/mol
MDL Number: MFCD06657887
CAS Number: 1043869-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR