Methyl 5-bromo-2-chlorothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A119222-1G A119222-250MG A119222-100MG
77009-336EA 95 USD
77009-336 77009-338 77889-821
Methyl 5-bromo-2-chlorothiazole-4-carboxylate 97%
Formula: C₅H₃BrClNO₂S
MDL Number: MFCD27922923
CAS Number: 1784255-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR