4-Chloro-N1-methylbenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A119180-100MG A119180-250MG A119180-5G A119180-1G
77009-324EA 18.04 USD
77009-324 77009-326 77009-328 77683-070
4-Chloro-N1-methylbenzene-1,2-diamine 97%
4-Chloro-N1-methylbenzene-1,2-diamine

Precaución: For research use only.

Formula: C₇H₉ClN₂
MW: 156.61 g/mol
MDL Number: MFCD10692801
CAS Number: 59681-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR