2-Phenylthiazole-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A119178-1G A119178-5G
77009-320EA 53.05 USD
77009-320 77009-322
2-Phenylthiazole-5-carbaldehyde 98%
2-Phenylthiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NOS
MW: 189.2300 g/mol
MDL Number: MFCD04973719
CAS Number: 1011-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR