2-Fluoro-6-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (6-Fluoro-o-tolyl)methanol, (2-Fluoro-6-methylphenyl)methanol

A119155-1G A119155-250MG
77009-316EA 29 USD
77009-316 77009-318
2-Fluoro-6-methylbenzyl alcohol 95%
2-Fluoro-6-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.155 g/mol
MDL Number: MFCD06660280
CAS Number: 478163-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR