(S)-Piperazine-2-carboxylic acid dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (2S)-piperazine-2-carboxylic acid dihydrochloride

A119140-10G A119140-1G A119140-250MG A119140-25G A119140-5G A119140-100G
76983-124EA 27 USD
76983-124 76983-126 76983-128 76983-130 76983-132 77889-817
(S)-Piperazine-2-carboxylic acid dihydrochloride 95%
(S)-Piperazine-2-carboxylic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂Cl₂N₂O₂
MDL Number: MFCD01632104
CAS Number: 158663-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR