(R)-2-((tert-Butoxycarbonyl)amino)propanoic acid hydrate 95%

Supplier: Ambeed
Warning

A118949-10G A118949-25G
76983-106EA 15.92 USD
76983-106 76983-108
(R)-2-((tert-Butoxycarbonyl)amino)propanoic acid hydrate 95%
(R)-2-((tert-Butoxycarbonyl)amino)propanoic acid hydrate

Precaución: for Research Use Only

Formula: C₈H₁₇NO₅
MW: 207.22 g/mol
MDL Number: MFCD26383924
CAS Number: 75638-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR