3,5-Difluoro-4-iodophenol 95%

Supplier: Ambeed
Warning

A118913-100MG A118913-250MG A118913-1G
77009-306EA 16 USD
77009-306 77009-308 77566-190
3,5-Difluoro-4-iodophenol 95%
3,5-Difluoro-4-iodophenol

Precaución: For research use only.

Formula: C₆H₃F₂IO
MDL Number: MFCD09955025
CAS Number: 1000574-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR