6-(Methoxymethyl)pyrimidin-4-ol 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A118793-100MG A118793-250MG A118793-5G A118793-1G
77009-298EA 19.1 USD
77009-298 77009-300 77009-302 77683-069
6-(Methoxymethyl)pyrimidin-4-ol 97%, Ambeed.Inc
6-(Methoxymethyl)pyrimidin-4-ol

Precaución: For research use only.

Formula: C₆H₈N₂O₂
MW: 140.13 g/mol
MDL Number: MFCD16630187
CAS Number: 3122-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR