4,5,6-Trichloropyrimidine 97%

Supplier: Ambeed
Warning

A118764-100G A118764-100MG A118764-1G A118764-250MG A118764-25G A118764-5G
76995-652EA 457 USD
76995-652 76995-654 76995-656 76995-658 76995-660 76995-662
4,5,6-Trichloropyrimidine 97%
4,5,6-Trichloropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₃N₂
MDL Number: MFCD08458457
CAS Number: 1780-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR