Methyl 3-bromo-2-(bromomethyl)-4,5-dimethoxybenzoate 95%

Supplier: Ambeed
Warning

A118597-100MG A118597-250MG
77009-288EA 94.43 USD
77009-288 77009-290
Methyl 3-bromo-2-(bromomethyl)-4,5-dimethoxybenzoate 95%
Methyl 3-bromo-2-(bromomethyl)-4,5-dimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂Br₂O₄
MW: 368.01 g/mol
MDL Number: MFCD25954782
CAS Number: 1042722-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR