1-(2-Methoxyphenoxy)-2,3-epoxypropane 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Epoxy-3-(2-methoxyphenoxy)propane, Glycidyl Guaiacol ether, 2-[(2-Methoxyphenoxy)methyl]oxirane, Glycidyl 2-methoxyphenyl ether, Glycidyl 2-Methoxyphenyl ether

A118533-100G A118533-10G A118533-1G A118533-250MG A118533-25G A118533-5G
76995-628EA 164.44 USD
76995-628 76995-630 76995-632 76995-634 76995-636 76995-638
1-(2-Methoxyphenoxy)-2,3-epoxypropane 95%
1-(2-Methoxyphenoxy)-2,3-epoxypropane

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
MDL Number: MFCD00190158
CAS Number: 2210-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR