3-Bromo-5-chlorothieno[3,2-b]pyridine 95%

Supplier: Ambeed
Warning

A118315-100MG A118315-1G A118315-250MG
76983-086EA 87 USD
76983-086 76983-088 76983-090
3-Bromo-5-chlorothieno[3,2-b]pyridine 95%
3-Bromo-5-chlorothieno[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₃BrClNS
MW: 248.52 g/mol
MDL Number: MFCD13176580
CAS Number: 912332-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR