2',3',5'-Tri-O-acetyl-2-fluoroadenosine 98%

Supplier: Ambeed
Warning

A118275-10G A118275-1G
76995-610EA 107 USD
76995-610 76995-612
2',3',5'-Tri-O-acetyl-2-fluoroadenosine 98%
2',3',5'-Tri-O-acetyl-2-fluoroadenosine

Precaución: for Research Use Only

Formula: C₁₆H₁₈FN₅O₇
MDL Number: MFCD08458339
CAS Number: 15811-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR