Vanillin 99%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

A118216-1000G A118216-100G A118216-10G A118216-25G A118216-500G
76995-600EA 49.87 USD
76995-600 76995-602 76995-604 76995-606 76995-608
Vanillin 99%
Vanillin

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00006942
CAS Number: 121-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR