tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate oxalate 95%

Supplier: Ambeed
Warning

A118195-1G A118195-250MG A118195-5G
76983-080EA 73.21 USD
76983-080 76983-082 76983-084
tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate oxalate 95%
tert-Butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₇
MW: 320.34 g/mol
MDL Number: MFCD27923424
CAS Number: 1588440-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR