(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A117879-100MG A117879-250MG
77026-226EA 134.74 USD
77026-226 77026-228
(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid 98%
(3S,4R)-1-tert-Butyl-4-(2,4-difluorophenyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉F₂NO₂
MW: 283.314 g/mol
MDL Number: MFCD13152278
CAS Number: 455957-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR