3-Amino-5-chloropyridine-2-carboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-5-chloropicolinic acid

A117680-100MG A117680-250MG
76983-066EA 50.93 USD
76983-066 76983-068
3-Amino-5-chloropyridine-2-carboxylic acid 96%
3-Amino-5-chloropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD09999771
CAS Number: 53636-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR