Methyl-2,6-dichloro-5-fluoronicotinate 97%

Supplier: Ambeed
Warning

A117540-100G A117540-1G A117540-250MG A117540-25G A117540-5G
76995-578EA 462 USD
76995-578 76995-580 76995-582 76995-584 76995-586
Methyl-2,6-dichloro-5-fluoronicotinate 97%
Methyl-2,6-dichloro-5-fluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₄Cl₂FNO₂
MDL Number: MFCD02180634
CAS Number: 189281-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR