Methyl 5-(aminomethyl)-2-chlorobenzoate hydrochloride 97%

Supplier: Ambeed
Warning

A117513-1G A117513-250MG A117513-5G
77026-212EA 59 USD
77026-212 77026-214 77026-216
Methyl 5-(aminomethyl)-2-chlorobenzoate hydrochloride 97%
Methyl 5-(aminomethyl)-2-chlorobenzoate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂NO₂
MW: 236.09 g/mol
MDL Number: MFCD16876981
CAS Number: 90942-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR