1-(Cyclopropylsulfonyl)piperidine-4-carbonitrile 97%

Supplier: Ambeed
Warning

A117455-1G A117455-250MG
76983-046EA 26 USD
76983-046 76983-048
1-(Cyclopropylsulfonyl)piperidine-4-carbonitrile 97%
1-(Cyclopropylsulfonyl)piperidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂S
MDL Number: MFCD19440953
CAS Number: 1036738-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR