Sorafenib tosylate (BAY 43-9006 tosylate salt) 98%

Supplier: Ambeed
Danger

A117299-100MG A117299-1G A117299-250MG A117299-25G A117299-50MG A117299-10G A117299-10MG
77026-198EA 8 USD
77026-198 77026-200 77026-202 77026-204 77026-206 77666-690 77666-692
Sorafenib tosylate (BAY 43-9006 tosylate salt) 98%
Sorafenib tosylate (BAY 43-9006 tosylate salt)

Precaución: for Research Use Only

Formula: C₂₁H₁₆ClF₃N₄O₃·C₇H₈O₃S
MDL Number: MFCD08235032
CAS Number: 475207-59-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR