tert-Butyl 3-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate 98%

Supplier: Ambeed
Warning

A117071-100MG A117071-1G A117071-250MG
77026-180EA 130.5 USD
77026-180 77026-182 77026-184
tert-Butyl 3-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate 98%
tert-Butyl 3-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂S
MW: 318.23 g/mol
MDL Number: MFCD09878671
CAS Number: 1000577-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR