2-Amino-5-chloro-3-nitropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine

A117037-1G A117037-250MG
77009-232EA 10 USD
77009-232 77009-234
2-Amino-5-chloro-3-nitropyridine 95%
2-Amino-5-chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.56
MDL Number: MFCD00092011
CAS Number: 5409-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR