GNF-5 98%

Supplier: Ambeed
Danger

A117001-100MG A117001-10MG A117001-1MG A117001-50MG
77026-164EA 117 USD
77026-164 77026-166 77026-168 77026-170
GNF-5 98%
GNF-5

Precaución: for Research Use Only

Formula: C₂₀H₁₇F₃N₄O₃
MDL Number: MFCD16877246
CAS Number: 778277-15-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR