Bis(2,4,6-trichlorophenyl)oxalate 98%

Supplier: Ambeed
Danger

Synonyms: Bis(2,4,6-trichlorophenyl) oxalate, TCPO, Oxalic Acid Bis(2,4,6-trichlorophenyl) Ester

A116994-100G A116994-10G A116994-1G A116994-250MG A116994-25G A116994-5G
76995-556EA 429.67 USD
76995-556 76995-558 76995-560 76995-562 76995-564 76995-566
Bis(2,4,6-trichlorophenyl)oxalate 98%
Bis(2,4,6-trichlorophenyl)oxalate

Precaución: for Research Use Only

Formula: C₁₄H₄Cl₆O₄
MW: 448.88 g/mol
MDL Number: MFCD00043061
CAS Number: 1165-91-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR