2-(Methoxycarbonyl)thiophene-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A116857-100MG
A116857-1G
A116857-250MG
76983-028EA
74.27
USD
76983-028
76983-030
76983-032
2-(Methoxycarbonyl)thiophene-3-carboxylic acid 98%
2-(Methoxycarbonyl)thiophene-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₆O₄S MW: 186.19 g/mol |
MDL Number:
MFCD20640613 CAS Number: 115777-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...