Methyl 4-formyl-3-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A116776-100MG A116776-1G A116776-250MG A116776-5G
76983-020EA 14 USD
76983-020 76983-022 76983-024 76983-026
Methyl 4-formyl-3-hydroxybenzoate 95%
Methyl 4-formyl-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈O₄
MDL Number: MFCD17015455
CAS Number: 24589-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR