2-(Allyloxy)phenol 97%
Supplier: Ambeed
|
Warning
|
A116664-250MG
A116664-25G
A116664-5G
77009-212EA
10.61
USD
77009-212
77009-214
77683-051
2-(Allyloxy)phenol 97%
2-(Allyloxy)phenol
Precaución: For research use only.
Formula:
C₉H₁₀O₂ MW: 150.175 g/mol |
MDL Number:
MFCD00072617 CAS Number: 1126-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...