2,6-Dibromo-4-methoxyaniline 98%

Supplier: Ambeed
Warning

A116652-1G A116652-250MG A116652-5G
76983-014EA 120 USD
76983-014 76983-016 77889-802
2,6-Dibromo-4-methoxyaniline 98%
2,6-Dibromo-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
MDL Number: MFCD06654495
CAS Number: 95970-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR