3-(Phenylmethoxy)-azetidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzyloxy)azetidine hydrochloride

A116617-1G A116617-250MG A116617-5G
77026-150EA 50 USD
77026-150 77026-152 77026-154
3-(Phenylmethoxy)-azetidine hydrochloride 97%
3-(Phenylmethoxy)-azetidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD09907646
CAS Number: 897019-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR