Ethyl pyridazine-3-carboxylate 97%

Supplier: Ambeed
Warning

A116450-100MG A116450-1G A116450-250MG
77026-132EA 39.26 USD
77026-132 77026-134 77026-136
Ethyl pyridazine-3-carboxylate 97%
Ethyl pyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD09908167
CAS Number: 1126-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR