(2,6-Difluoro-3,5-dimethoxyphenyl)methanol 97%

Supplier: Ambeed
Warning

A116417-100MG A116417-10G A116417-1G A116417-250MG A116417-5G
76982-990EA 50.93 USD
76982-990 76982-992 76982-994 76982-996 76982-998
(2,6-Difluoro-3,5-dimethoxyphenyl)methanol 97%
(2,6-Difluoro-3,5-dimethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₃
MW: 204.17 g/mol
MDL Number: MFCD22683867
CAS Number: 1208434-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR