6-Bromo-2-chloro-4-methylquinoline 95%

Supplier: Ambeed
Warning

A116378-100MG A116378-1G A116378-250MG
77026-126EA 19.1 USD
77026-126 77026-128 77026-130
6-Bromo-2-chloro-4-methylquinoline 95%
6-Bromo-2-chloro-4-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₇BrClN
MW: 256.5300 g/mol
MDL Number: MFCD06254262
CAS Number: 3913-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR