3-Amino-3-hydroxymethyloxetane 95%

Supplier: Ambeed
Warning

Synonyms: (3-Aminooxetan-3-yl)methanol

A116312-100MG A116312-1G A116312-250MG A116312-5G
77009-196EA 77 USD
77009-196 77009-198 77009-200 77009-202
3-Amino-3-hydroxymethyloxetane 95%
3-Amino-3-hydroxymethyloxetane

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MDL Number: MFCD20926150
CAS Number: 1305208-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR