4-Fluoro-1H-indole-3-carbonitrile 97%

Supplier: Ambeed
Warning

A116237-1G A116237-250MG
76982-984EA 310.85 USD
76982-984 76982-986
4-Fluoro-1H-indole-3-carbonitrile 97%
4-Fluoro-1H-indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅FN₂
MW: 160.148 g/mol
MDL Number: MFCD15527152
CAS Number: 1260759-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR