Methyl 1,3-benzodioxole-4-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl benzo[d][1,3]dioxole-4-carboxylate

A116191-1G A116191-5G
76995-530EA 26.53 USD
76995-530 76995-532
Methyl 1,3-benzodioxole-4-carboxylate 98%
Methyl 1,3-benzodioxole-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.157 g/mol
MDL Number: MFCD01546796
CAS Number: 33842-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR