5-Bromo-2-fluoro-4-methylbenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-fluoro-4-methylphenylboronic acid

A116158-1G A116158-250MG A116158-5G
77026-112EA 14 USD
77026-112 77026-114 77026-116
5-Bromo-2-fluoro-4-methylbenzeneboronic acid 98%
5-Bromo-2-fluoro-4-methylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
MDL Number: MFCD09258744
CAS Number: 957061-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR