1-(5-(Benzyloxy)-2-hydroxyphenyl)ethanone 95%

Supplier: Ambeed
Warning

A116145-1G A116145-250MG
76982-978EA 101.85 USD
76982-978 76982-980
1-(5-(Benzyloxy)-2-hydroxyphenyl)ethanone 95%
1-(5-(Benzyloxy)-2-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00598842
CAS Number: 30992-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR