4,6-Dichloro-3-methyl-1H-pyrazolo[3,4-d]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A116124-100MG
A116124-250MG
A116124-5G
77026-108EA
84.88
USD
77026-108
77026-110
77570-984
4,6-Dichloro-3-methyl-1H-pyrazolo[3,4-d]pyrimidine 97%
4,6-Dichloro-3-methyl-1H-pyrazolo[3,4-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₄Cl₂N₄ MW: 203.0288 g/mol |
MDL Number:
MFCD16249844 CAS Number: 1211522-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...