5-Nitro-2-piperidinoaniline 95%

Supplier: Ambeed
Warning

A116055-250MG A116055-5G A116055-1G
76995-526EA 14 USD
76995-526 76995-528 77880-937
5-Nitro-2-piperidinoaniline 95%
5-Nitro-2-piperidinoaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD00852821
CAS Number: 5367-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR