2-Bromobenzene-1,3,5-triol 95%

Supplier: Ambeed
Warning

A116029-1G A116029-250MG A116029-5G
76982-962EA 66 USD
76982-962 76982-964 77885-060
2-Bromobenzene-1,3,5-triol 95%
2-Bromobenzene-1,3,5-triol

Precaución: for Research Use Only

Formula: C₆H₅BrO₃
MDL Number: MFCD22056138
CAS Number: 84743-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR