(5-(4-Fluorophenyl)thiophen-2-yl)(5-iodo-2-methylphenyl)methanone 95%

Supplier: Ambeed
Warning

A115925-25G A115925-5G
77009-166EA 269.47 USD
77009-166 77009-168
(5-(4-Fluorophenyl)thiophen-2-yl)(5-iodo-2-methylphenyl)methanone 95%
(5-(4-Fluorophenyl)thiophen-2-yl)(5-iodo-2-methylphenyl)methanone

Precaución: for Research Use Only

Formula: C₁₈H₁₂FIOS
MW: 422.26 g/mol
MDL Number: MFCD12405594
CAS Number: 1071929-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR