1-Amino-7-bromoisoquinoline 97%

Supplier: Ambeed
Warning

A115885-100MG A115885-10G A115885-1G A115885-250MG A115885-5G
77009-156EA 30 USD
77009-156 77009-158 77009-160 77009-162 77009-164
1-Amino-7-bromoisoquinoline 97%
1-Amino-7-bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD07374397
CAS Number: 215453-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR